Learn More
Palladium(II) sulfate dihydrate, Premion™, 99.995% (metals basis), Thermo Scientific Chemicals
189.95 € - 864.30 €
Chemical Identifiers
CAS | 13444-98-9 |
---|---|
Molecular Formula | H4O6PdS |
Molecular Weight (g/mol) | 238.51 |
MDL Number | MFCD00011173 |
InChI Key | TWIRRPLUAGEFNJ-UHFFFAOYSA-L |
Synonym | palladium ii sulfate dihydrate, palladium ii sulfate dihydrate, premion, palladium sulfate dihydrate, so4.pd.2h2o, palladium 2+ sulfate dihydrate, palladium 2+ dihydrate sulfate, palladium 2+ sulfate-water 1/1/2, palladium 2+ ion dihydrate sulfate, palladium ii sulfate dihydrate metals basis |
PubChem CID | 22042048 |
IUPAC Name | palladium(2+);sulfate;dihydrate |
SMILES | O.O.[Pd++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11340288
|
Thermo Scientific Alfa Aesar
043698.03 |
1 g |
189.95 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11330288
|
Thermo Scientific Alfa Aesar
043698.06 |
5 g |
864.30 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13444-98-9 | |
238.51 | |
TWIRRPLUAGEFNJ-UHFFFAOYSA-L | |
22042048 | |
O.O.[Pd++].[O-]S([O-])(=O)=O |
H4O6PdS | |
MFCD00011173 | |
palladium ii sulfate dihydrate, palladium ii sulfate dihydrate, premion, palladium sulfate dihydrate, so4.pd.2h2o, palladium 2+ sulfate dihydrate, palladium 2+ dihydrate sulfate, palladium 2+ sulfate-water 1/1/2, palladium 2+ ion dihydrate sulfate, palladium ii sulfate dihydrate metals basis | |
palladium(2+);sulfate;dihydrate |
Specifications
(decomposition) | |
Brown to Red | |
H4O6PdS | |
MFCD00011173 | |
palladium ii sulfate dihydrate, palladium ii sulfate dihydrate, premion, palladium sulfate dihydrate, so4.pd.2h2o, palladium 2+ sulfate dihydrate, palladium 2+ dihydrate sulfate, palladium 2+ sulfate-water 1/1/2, palladium 2+ ion dihydrate sulfate, palladium ii sulfate dihydrate metals basis | |
O.O.[Pd++].[O-]S([O-])(=O)=O | |
238.51 | |
238.51 (202.48 Anhydrous) | |
Premion™ | |
(metals basis) |
13444-98-9 | |
Powder | |
PdSO4·2H2O | |
UN3260 | |
TWIRRPLUAGEFNJ-UHFFFAOYSA-L | |
palladium(2+);sulfate;dihydrate | |
22042048 | |
99.995% | |
Hygroscopic | |
Palladium (II) sulfate dihydrate |
Safety and Handling
GHS H Statement
H314-H318-H290-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.
P234-P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H302-H314-H334-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 236-957-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only