missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) sulfide, 99% (metals basis), Pd 72% min, Thermo Scientific Chemicals

242.00 € - 1164.00 €

Chemical Identifiers

CAS 12125-22-3
Molecular Formula PdS
Molecular Weight (g/mol) 138.48
MDL Number MFCD00016285
InChI Key FEWBURNPAYHODO-UHFFFAOYSA-N
Synonym palladium ii sulfide, palladium sulphide, palladium monosulphide, palladium sulfide, acmc-20ajzj, palladium sulfide,, palladium ii sulfide trace metals basis
PubChem CID 82926
SMILES [S--].[Pd++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11307507
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
012674.03
1 g
242.00 €
1g
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
11397497
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Alfa Aesar
012674.06
5 g
1164.00 €
5g
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in scientific research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12125-22-3
138.48
FEWBURNPAYHODO-UHFFFAOYSA-N
82926
PdS
MFCD00016285
palladium ii sulfide, palladium sulphide, palladium monosulphide, palladium sulfide, acmc-20ajzj, palladium sulfide,, palladium ii sulfide trace metals basis
[S--].[Pd++]

Specifications

950°C (decomposition)
Crystalline
MFCD00016285
Insoluble in water.
[S--].[Pd++]
138.48
138.46
Pd 72% min.
6.60 g/mL
Palladium(II) sulfide
12125-22-3
PdS
palladium ii sulfide, palladium sulphide, palladium monosulphide, palladium sulfide, acmc-20ajzj, palladium sulfide,, palladium ii sulfide trace metals basis
FEWBURNPAYHODO-UHFFFAOYSA-N
sulfanylidenepalladium
82926
99%
Sulphurous
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 235-190-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only