missing translation for 'onlineSavingsMsg'
Learn More

Palmitoyl chloride, 98%, Thermo Scientific Chemicals

28.60 € - 659.28 €

Chemical Identifiers

CAS 112-67-4
MDL Number MFCD00000742
InChI Key ARBOVOVUTSQWSS-UHFFFAOYSA-N
Synonym palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride
PubChem CID 8206
IUPAC Name hexadecanoyl chloride
SMILES CCCCCCCCCCCCCCCC(=O)Cl
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10731161
View Documents
Thermo Scientific Acros
304290050
5 mL Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10668021
View Documents
Thermo Scientific Acros
304291000
100 mL Glass bottle
75.58 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10013690
View Documents
Thermo Scientific Acros
304295000
500 mL Glass bottle
237.18 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10217463
View Documents
Thermo Scientific Acros
304290025
2.5 L Glass bottle
659.28 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112-67-4
ARBOVOVUTSQWSS-UHFFFAOYSA-N
8206
CCCCCCCCCCCCCCCC(=O)Cl
MFCD00000742
palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride
hexadecanoyl chloride

Specifications

1% max. (as palmitic acid)
0.9000g/mL
98%
CH3(CH2)14COCl
MFCD00000742
palmitoyl chloride, palmitic acid chloride, palmitoylchloride, unii-7642r2josb, hexadecanoyl-chloride, palm acetyl chlorine 14 acetyl chlorine, hexadecanoic acid, chloride, palmitic chloride, hexadecanoylchloride, n-hexadecanoyl chloride
CCCCCCCCCCCCCCCC(=O)Cl
274.873
274.88
Liquid
112-67-4
Authentic
C16H31ClO
1.4510 to 1.4530 (20°C, 589nm)
02, 374
ARBOVOVUTSQWSS-UHFFFAOYSA-N
hexadecanoyl chloride
8206
98 to 101% (Argentometry), ≥97.5%(GC)
Palmitoyl chloride, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store in a dry place. Store in a closed container.

Danger

missing translation for 'einecsNumber' : 203-996-7

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only