missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Panoramasque™ Full-Face Air Purifying Respirator (APR)

The Panoramasque™ Full-Face APR is a Mono filter with Rd40 thread connection system that offers respiratory protection, depending on the canister type assembled on it.

Brand:  Honeywell™ 1710980

 View more versions of this product

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15952912

  • 260.00 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Product Type: Full-Face Air Purifying Respirator (APR)
  • Material (Body): EPDM or Silicone
  • Material (Lens): PMMA, Glass, Chlorine Hazard Specific
  • Color: Black, Yellow
  • Cartridge: Single Filter via Rd40 (1/7 in.) Connection
  • Use with: EN141 or EN143 Class 1 or 2 Cert. Honeywell Filters
  • PPE Class: Variable (Dependant on Cartridge)
  • Hazard Types: Particulates, Gas, Vapor, Smoke
  • Applications: Chemical Industry, Oil and Gas Industries, Pharmaceutical Industry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EN 136:1989
Yellow
Full-Face Air Purifying Respirator (APR)
Chemical Industry, Oil and Gas Industries, Pharmaceutical Industry
Five-Point Harness
PMMA
1/7 in. Rd40 Thread System
Mono-Filter
Silicone
EN141 or EN143 Class 1 or 2 Cert. Honeywell Filters
Without Speech Diaphragm
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.