missing translation for 'onlineSavingsMsg'
Learn More

Paraffin, liquid, pure, Thermo Scientific Chemicals

25.02 € - 101.51 €

Chemical Identifiers

CAS 8012-95-1
Molecular Formula MFCD00131611
Molecular Weight (g/mol) 0.00
MDL Number MFCD00131611
InChI Key FFNDMZIBVDSQFI-UHFFFAOYSA-N
Synonym delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
PubChem CID 68245
ChEBI CHEBI:38701
IUPAC Name 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
SMILES *
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Prijs Hoeveelheid en beschikbaarheid  
10689014
missing translation for 'viewDocuments'
Thermo Scientific Acros
171402500
250 mL Glass bottle
25.02 €
250mL
Please to purchase this item. Need a web account? Register with us today!
10361124
missing translation for 'viewDocuments'
Thermo Scientific Acros
171400010
1 L Glass bottle
54.87 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10540614
missing translation for 'viewDocuments'
Thermo Scientific Acros
171400025
2.5 L Glass bottle
101.51 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Beschrijving

Beschrijving

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specificatie

Chemische identificatiekenmerken

8012-95-1
0.00
FFNDMZIBVDSQFI-UHFFFAOYSA-N
68245
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
MFCD00131611
MFCD00131611
delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
CHEBI:38701
*

Specificatie

8012-95-1
Colorless
300.0°C
Authentic
MFCD00131611
0.88
Solubility in water: insoluble
*
0.00
CHEBI:38701
Pure
Paraffin, liquid, Pure
-24.0°C
0.8800g/mL
218°C
MFCD00131611
14, 7186
delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
FFNDMZIBVDSQFI-UHFFFAOYSA-N
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
68245
65-75 mm2/s (40°C)
Oily Liquid
Video's
Veiligheid en behandeling

Veiligheid en behandeling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 232-384-2

Productidentificatie
  • Paraffin, liquid

Signaalwoord
  • Warning

Gevarencategorie
  • Serious eye damage/eye irritation Category 2

Gevarenaanduiding
  • H319-Causes serious eye irritation.

Veiligheidsmaatregel
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documentatie

Documentatie

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.