missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bemis™ Parafilm™ M Laboratory Wrapping Film
Efficient and cost-effective covering for test tubes, beakers, vials, petri dishes, flasks, and various other instruments and objects with irregular surfaces.
40.45 € - 850.41 €
Specifications
Applications | Efficient and cost-effective covering, over wrap to further secure stoppers, lids, and caps, and shelf or tray liner to prevent slipping of bottles or instruments. |
---|---|
Product Type | Sealing Film |
Material | Thermoplastic |
Product Code | Brand | Width (English) | Length (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (English) | Length (English) | Price | Quantity & Availability | ||||
11747487
|
Bemis™
HS120265 |
20 in. | 50 ft. |
101.00 €
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||||
10018130
|
Bemis™
HS234526C |
4 in. | 250 ft. |
80.80 €
Each |
In Stock
Log in to see stock. |
||||
11693119
|
Bemis™
PM992 |
2 in. | 250 ft. | N/A | |||||
11762644
|
Bemis™
HS234526A |
2 in. | 250 ft. |
41.95 €
Each |
In Stock
Log in to see stock. |
||||
11772644
|
Bemis™
HS234526B |
4 in. | 125 ft. |
40.45 €
Each |
In Stock
Log in to see stock. |
||||
Description
Efficient and cost-effective covering for test tubes, beakers, vials, petri dishes, flasks, and various other instruments and objects with irregular surfaces.
- Protect the integrity of research applications with a tight moisture-barrier that holds a strong seal and doesn’t release from glassware, despite off-gassing
- Reduce the risk of tampered data with a covering that is odorless, colorless and semi-transparent
- Prevents moisture and volume loss due to material composition and tight seal
- Flexible and self-sealing material allows it to cling around irregular shapes and surfaces
- Stretches more than 200% of original length
- Resistant up to 48 hours against many polar substances such as saline solutions, inorganic acids and alkaline solutions
Permeability
- Oxygen: (ASTM 1927-98) 150 cc/m² d at 23°C and 50% RH
- Carbon Dioxide: (Modulated IR Method) 1200 cc/m² d at 23°C and 0% RH
- Water Vapor: (ASTM F1249-01)
- Flat: 1 g/m² d at 38°C and 90% RH
- Creased: 1 g/m² d at 38°C and 90% RH
Effects of Common Reagents
- Potassium Permanganate
- 5%: No apparent effect except permanent dark brown coloration in 18 hours
- 0.1%: Same as 5% except slightly less color
- Iodine solution (0.1N)
- No effect except staining brown in 18 hours
- Ethyl Alcohol (95%)
- No apparent effect except some face whitening in 24 hours
- No apparent effect in 24 hours:
- Hydrochloric Acid conc. (12N) dil. (5N)
- Sulphuric Acid conc. (36N) dil. (5N)
- Nitric Acid conc. (16N) dil. (5N)
- Sodium Hydroxide conc. (22%)
- Ammonium Hydroxide conc. (28% NH<sub>3</sub>)
- Salt (NaCl) solution (20%)
- Isopropyl Alcohol (99%)
- Not recommended for use with chlorinated, non-polar aliphatic and aromatic solvents
- Film becomes soft and sticky at about 130° to 150°F (54° to 66°C)
Order Info
13-374-10, 13-374-12, and 13-374-16 rolls are packaged in cardboard dispensing cartons.
Specifications
Efficient and cost-effective covering, over wrap to further secure stoppers, lids, and caps, and shelf or tray liner to prevent slipping of bottles or instruments. | |
Sealing Film | |
Thermoplastic |