Learn More
Amcor Parafilm™ M Laboratory Film
Efficient and cost-effective covering for test tubes, beakers, vials, petri dishes, flasks, and various other instruments and objects with irregular surfaces.
Brand:  Amcor 120265
 View more versions of this product
Product Code. 11747487
  • 101.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Show More
Specifications
Specifications
Efficient and cost-effective covering, over wrap to further secure stoppers, lids, and caps, and shelf or tray liner to prevent slipping of bottles or instruments.
Natural
Sealing Film
51 cm
Thermoplastic
50 ft.
Amcor PM998
20 in.
15.2 m
Parafilm Wrap
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Amcor Parafilm™ M Laboratory Film > Roll W x L: 20 in. x 50 ft. (51cm x 15.2m)