missing translation for 'onlineSavingsMsg'
Learn More
Paraformaldehyde, 96%, Thermo Scientific Chemicals Product Code.: 11473704

Paraformaldehyde, 96%, Thermo Scientific Chemicals

Product Code. 11473704
500 g
Click to view available options
Quantity:
10,000 g
100 g
2500 g
500 g
This item is not returnable. View return policy

Product Code. 11473704

Brand: Thermo Scientific Alfa Aesar A11313.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Paraformaldehyde is used as a fumigant, a disinfectant and in the manufacture of synthetic resins like melamine resin, phenol resin and polyacetal resin. It is also used in cell culture. It also serves as a fixative in electron microscopy. It is also used in the preparation of formalin fixatives for tissues or cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula CH2O
Molecular Weight (g/mol) 30.026
MDL Number MFCD00133991
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehydeShow MoreShow Less
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Melting Point 120°C to 170°C (decomposition)
Density 1.3
Flash Point 71°C (159°F)
Quantity 500 g
UN Number UN2213
Beilstein 102769
Merck Index 14,7025
Solubility Information Soluble in hot water. Insoluble in water,ether and alcohol.
Percent Purity 97%
Chemical Name or Material Paraformaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252586 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog NumberA11313.0C,A11313.36,A11313.0E,A11313.22
Lot Number10250778 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA11313.0C,A11313.36,A11313.0E,A11313.22
Lot Number10240367 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberA11313.0C,A11313.36,A11313.0E,A11313.22
Lot Number10248852 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberA11313.0C,A11313.36,A11313.0E,A11313.22
Lot Number10247775 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog NumberA11313.0C,A11313.36,A11313.0E,A11313.22
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Paraformaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Paraformaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Paraformaldehyde, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.