Learn More
Paraformaldehyde, 96%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Decomposition Information | 260°C |
Melting Point | 120.0°C to 170.0°C |
Color | White |
Density | 1.03 (Air = 1.0) |
pH | 4.0 to 5.5 (10% suspension) |
Flash Point | 71°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% |
Packaging | Plastic bottle |
Linear Formula | (CH2O)n |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468180 | Certificate of Analysis | 31/01/2025 | 416780250,416780030,416785000,416780100,416780010 |
A0467184 | Certificate of Analysis | 28/10/2024 | 416780030,416780250,416785000,416780100,416780010 |
A0466231 | Certificate of Analysis | 25/09/2024 | 416780030,416780250,416785000,416780100,416780010 |
A0465707 | Certificate of Analysis | 27/08/2024 | 416780030,416780250,416785000,416780100,416780010 |
B0155763 | Certificate of Analysis | 11/06/2024 | 416780030,416780250,416785000,416780100,416780010 |
Safety and Handling




- Paraformaldehyde
- Danger
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Carcinogenicity Category 2
- Acute toxicity Category 4
- Flammable solid Category 2
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.