missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Pararosaniline chloride, 96%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ideal for analysis by ICP-MS and high-resolution ICP-MS.
- Evaluated for 65 metals at low-level ppt concentrations.
- Manufactured, purified, and packaged in a Class 1000 clean room.
- Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
- Three year shelf life from time of manufacture - available on COA

Specifications
Specifications
Melting Point | 268.0°C to 270.0°C |
Color | Green |
Physical Form | Crystalline Powder |
Quantity | 5 g |
Assay Percent Range | 97.5% min. (HPLC) |
Solubility Information | Solubility in water: 10g/L (25°C). Other solubilities: 65g/L ethanol |
Formula Weight | 323.83 |
Percent Purity | 95% |
Grade | Pure |
Infrared Spectrum | Authentic |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0453205 | Certificate of Analysis | 25/08/2023 | 416800250,416800050,416801000 |
A0396964 | Certificate of Analysis | 14/12/2022 | 416800250,416800050,416801000 |
A0413347 | Certificate of Analysis | 13/12/2022 | 416800250,416800050,416801000 |
A0445778 | Certificate of Analysis | 26/09/2022 | 416800250,416800050,416801000 |
A0430168 | Certificate of Analysis | 20/05/2021 | 416800250,416800050,416801000 |
Safety and Handling

Product Identifier
- Pararosaniline chloride
- Danger
- Carcinogenicity Category 1B
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals Pararosaniline chloride, 96%, pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction