Learn More
Emma Safety Footwear Paris Safety Shoes
Lightweight woven, polyurethane-coated textile with Nano Carbon Composite protective toe. Performs excellently at workplaces where safety and long-lasting comfort are top priorities. The extremely comfortable PU insole guarantees long-lasting support and cushioning of the foot.
Brand:  Emma Safety Footwear MM40464746
 View more versions of this product
Product Code. 15928411
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • S1P-Safety shoe with sole plate for dry work
  • Woven Textile
  • Polyurethane coated
  • Hydro-Tec™ Antibacterial
  • Metal-free
  • Chrome-free lining
  • ESD certified
Specifications
Specifications
EN ISO 20345:2011
Black, Gray
Hydro-Tec™ Antibacterial
46
Laces
Paris
Textile
Composite