missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emma Safety Footwear Paris Safety Shoes
Lightweight woven, polyurethane-coated textile with Nano Carbon Composite protective toe. Performs excellently at workplaces where safety and long-lasting comfort are top priorities. The extremely comfortable PU insole guarantees long-lasting support and cushioning of the foot.
95.03 € - 130.00 €
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Closure Type | Laces |
Color | Black, Gray |
Design | Paris |
Liner Material | Hydro-Tec™ Antibacterial |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15958401
|
Emma Safety Footwear
MM40464739 |
39 |
111.00 €
1 pair |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
15968401
|
Emma Safety Footwear
MM40464740 |
40 |
111.00 €
1 pair |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
15978401
|
Emma Safety Footwear
MM40464741 |
41 |
127.00 €
1 pair |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
15988401
|
Emma Safety Footwear
MM40464742 |
42 |
111.00 €
1 pair |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
15998401
|
Emma Safety Footwear
MM40464743 |
43 |
130.00 €
1 pair |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
15908411
|
Emma Safety Footwear
MM40464744 |
44 |
111.00 €
1 pair |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
15918411
|
Emma Safety Footwear
MM40464745 |
45 |
111.00 €
1 pair |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
15928411
|
Emma Safety Footwear
MM40464746 |
46 | N/A | |||||||
15938411
|
Emma Safety Footwear
MM40464747 |
47 |
111.00 €
1 pair |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
Description
- S1P-Safety shoe with sole plate for dry work
- Woven Textile
- Polyurethane coated
- Hydro-Tec™ Antibacterial
- Metal-free
- Chrome-free lining
- ESD certified
Specifications
EN ISO 20345:2011 | |
Black, Gray | |
Hydro-Tec™ Antibacterial | |
Composite |
Laces | |
Paris | |
Textile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Emma Safety Footwear Paris Safety Shoes