Learn More
Emma Safety Footwear Paris Safety Shoes
Lightweight woven, polyurethane-coated textile with Nano Carbon Composite protective toe. Performs excellently at workplaces where safety and long-lasting comfort are top priorities. The extremely comfortable PU insole guarantees long-lasting support and cushioning of the foot.
Brand:  Emma Safety Footwear MM40464739
 View more versions of this product
Product Code. 15958401
  • 104.68 € / 1 pair
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S1P-Safety shoe with sole plate for dry work
  • Woven Textile
  • Polyurethane coated
  • Hydro-Tec™ Antibacterial
  • Metal-free
  • Chrome-free lining
  • ESD certified
Specifications
Specifications
EN ISO 20345:2011
Black, Gray
Hydro-Tec™ Antibacterial
39
Laces
Paris
Textile
Composite