Learn More
Emma Safety Footwear Paris Safety Shoes
Lightweight woven, polyurethane-coated textile with Nano Carbon Composite protective toe. Performs excellently at workplaces where safety and long-lasting comfort are top priorities. The extremely comfortable PU insole guarantees long-lasting support and cushioning of the foot.
Brand: Emma Safety Footwear MM40464741
This item is not returnable.
View return policy
Description
- S1P-Safety shoe with sole plate for dry work
- Woven Textile
- Polyurethane coated
- Hydro-Tec™ Antibacterial
- Metal-free
- Chrome-free lining
- ESD certified
Specifications
EN ISO 20345:2011 | |
Black, Gray | |
Hydro-Tec™ Antibacterial | |
41 |
Laces | |
Paris | |
Textile | |
Composite |