missing translation for 'onlineSavingsMsg'
Learn More

KARTELL Pasteur Pipette

These disposable Pasteur Pipettes are ideal for the precise transfer of samples.

Brand:  KARTELL 00329/1

 View more versions of this product

Product Code. 17848085

  • 89.70 € / Pack of 500
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Disposable PE pipette with bulb and graduations.
  • The slender tip guarantees precision transfer of the sample.
  • Sterile pipettes individually wrapped.
Specifications

Specifications

3.5 mL
Sterile
No
4 mL
Pasteur Pipet
Sterile
Polyethylene
28
Clear
150 mm
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.