missing translation for 'onlineSavingsMsg'
Learn More

Intelsius™ PathoPak

PathoPak sample transport packaging is certified for Category A and B shipments, meeting IATA, ADR, and CFR 49 (DOT) transport regulations, providing protection for your samples regardless of their specimen classification.

Encompass_Preferred

POA

Specifications

Certifications/Compliance UN3373
Type Sample Transport Packaging
View More Specs

Products 2
Product Code Brand Capacity Payload Price Quantity & Availability  
Product Code Brand Capacity Payload Price Quantity & Availability  
15645609
Encompass_Preferred
View Documents
Intelsius™
PP011
1 L 90 x 170 mm (3.5 x 6.7 in.)
POA
Pack of 20
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

12944007
Encompass_Preferred
View Documents
Intelsius™
PP013
3 L 125 x 255 mm (4.9 x 10.0 in.)
POA
Pack of 10
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

  • A Strong, durable and highly protective shipping solution for UN3373 category B biological samples.
  • Suitable for shipping a wide range of primaries including:
    • Blood Tubes
    • Specimen Containers
    • Swabs
  • Components Supplied:
    • 95kPa IATA compliant secondary rigid pressure vessel
    • Outer carton printed in compliance with transport regulations
    • Absorbent
    • Bubble wrap
    • Infectious class label
    • Security Seal label
Specifications

Specifications

UN3373
Sample Transport Packaging
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Intelsius™ PathoPak

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.