Learn More
Intelsius™ PathoShield™
The PathoShield™ is a complete shipping solution designed for the transportation of Category A and Category B biological materials. Each system is pre-printed with the required markings and includes all the components required to transport your sample safely.
Brand:  Intelsius™ PA002
Encompass_Preferred
 View more versions of this product
Product Code. 17368599
  • POA / Pack of 25
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • Complete Category B shipping solution includes flexible 95kPa tested compliant secondary, absorbent, labels, and relevant UN markings
  • Range of sizes suitable for shipping through postal and courier networks
  • Suitable for a wide range of primaries including blood vials, swabs and sample containers
  • Supplied flat packed for efficient storage and shipping prior to use
Specifications
Specifications
Pouch
Sample Transport Packaging
291 x 191 x 102 mm (11.4 x 7.5 x 4.0 in.)
4
Pouch: 228 x 250 mm (8.9 x 9.8 in.)