Learn More
Intelsius™ PathoShield™
The PathoShield™ is a complete shipping solution designed for the transportation of Category A and Category B biological materials. Each system is pre-printed with the required markings and includes all the components required to transport your sample safely.
Encompass_Preferred
POA - 195.45 €
Specifications
Includes Pouch
Type Sample Transport Packaging
View More Specs
Products 3
Product Code Brand Size Dimensions (LxWxH) Price Quantity & Availability  
Product Code Brand Size Dimensions (LxWxH) Price Quantity & Availability  
17378599
Encompass_Preferred
View Documents
Intelsius™
PA028
8 191 x 145 x 22 mm (7.5 x 5.7 x 0.9 in.) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17368599
Encompass_Preferred
View Documents
Intelsius™
PA002
4 291 x 191 x 102 mm (11.4 x 7.5 x 4.0 in.)
POA
Pack of 25
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15681632
Encompass_Preferred
View Documents
Intelsius™
PA025
5 275 x 179 x 43 mm (10.8 x 7.0 x 1.7 in.)
195.45 €
Pack of 25
View alternative products
Save Up to 
»
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
Description
Description
  • Complete Category B shipping solution includes flexible 95kPa tested compliant secondary, absorbent, labels, and relevant UN markings
  • Range of sizes suitable for shipping through postal and courier networks
  • Suitable for a wide range of primaries including blood vials, swabs and sample containers
  • Supplied flat packed for efficient storage and shipping prior to use
Specifications
Specifications
Pouch
Sample Transport Packaging
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Intelsius™ PathoShield™