missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS, pH 7.2
GREENER_CHOICE

PBS, pH 7.2

26.77 € - 238.00 €

Specifications

Additive No Sodium Pyruvate
Content And Storage 15°to 30°C
Dilution 1 X
Product Type Phosphate Buffered Saline (PBS)
For Use With (Application) Mammalian Cell Culture
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11530546
GREENER_CHOICE
View Documents
Gibco™
20012019
500mL
26.77 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11540546
GREENER_CHOICE
View Documents
Gibco™
20012068
10 x 500 mL
238.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation.

We offer a variety of Gibco™ PBS formulations for a range of cell culture applications.

cGMP Manufacturing and Quality System
Gibco™ PBS is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical Gibco™ PBS product made in our Scotland facility (70011-036). This facility is also registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
Specifications

Specifications

No Sodium Pyruvate
1 X
Mammalian Cell Culture
7.2
15°to 30°C
Phosphate Buffered Saline (PBS)
280 to 320 mOsm/kg
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 36 Month(s)

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ PBS, pH 7.2

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.