missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS, pH 7.2
GREENER_CHOICE

PBS, pH 7.2

Brand:  Gibco™ 20012019

14.40 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.


 View more versions of this product

Product Code. 11530546

  • 26.77 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Phosphate-buffered saline (PBS) is a balanced salt solution that is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation.

We offer a variety of Gibco™ PBS formulations for a range of cell culture applications. The complete formulation is available.

cGMP Manufacturing and Quality System
Gibco™ PBS is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical Gibco™ PBS product made in our Scotland facility (70011-036). This facility is also registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

No Sodium Pyruvate
15°to 30°C
Phosphate Buffered Saline (PBS)
Mammalian Cell Culture
7.2
500 mL
1 X
500mL
280 to 320 mOsm/kg
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ PBS, pH 7.2 > 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.