missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS, pH 7.4
GREENER_CHOICE

PBS (phosphate buffered saline) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

Brand:  Gibco™ 10010031

47.22 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.


 View more versions of this product

Product Code. 10728775

  • 50.50 € / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

PBS (phosphate buffered saline) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. PBS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation. We offer a variety of PBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

This PBS is manufactured as follows:

Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

No Sodium Pyruvate
Clear
1 X
Phosphate Buffered Saline (PBS)
Phosphate Buffered Saline
280 - 315 mOsm/kg
No Phenol Red
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
Animal Origin-Free
Storage conditions: 15 – 30°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Gibco™
1000 mL
Washing Cells Before Dissociation; Diluting Cells for Counting and Preparing Reagents; Transporting Cells or Tissue
7.4
Room Temperature
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2572130 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number10010002,10010001,10010023,10010031
Lot Number2494223 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number10010002,10010001,10010023,10010031
Lot Number2494226 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number10010002,10010001,10010023,10010031
Lot Number2562446 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number10010002,10010001,10010023,10010031
Lot Number2436334 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number10010002,10010001,10010023,10010031
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ PBS, pH 7.4 > Quantity: 1000 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.