missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PBS, pH 7.4 (flow cytometry grade)

Phosphate-Buffered Saline (PBS) is a balanced salt solution used for a variety of applications, including reagent preparation, diluting cells for flow cytometry and as a cell culture reagent.

Brand:  Gibco™ A1286301

46.09 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


Product Code. 12083469

  • 50.50 € / 10 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Phosphate-Buffered Saline (PBS) is a balanced salt solution used for a variety of applications, including reagent preparation, diluting cells for flow cytometry and as a cell culture reagent. This Gibco™ PBS is formulated without calcium and magnesium for suspending cells for flow cytometry. This PBS is not intended for cell culture as it is not tested for sterility. We offer a variety of Gibco™ PBS formulations for cell culture applications. Find the right cell culture product using the media selector tool.

This PBS is manufactured as follows:
Without: Calcium, Magnesium, Phenol Red

Product Intended Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

No Calcium, No Magnesium
1 X
10 L
7.4
Room Temperature
Liquid
Storage conditions: 15–30°C
Shipping conditions: Ambient
Shelf life: 9 months from date of manufacture
Phosphate Buffered Saline (PBS)
Phosphate Buffered Saline
No Phenol Red
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ PBS, pH 7.4 (flow cytometry grade) > 10 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.