Learn More
BRAND™ PCR 12-Strip Tubes
Manufactured from polypropylene without lubricants or chemical additives that can contaminate reagents and samples.PCR 12-Strip Tubes are autoclavable and RNase-, DNase-, and DNA-free. Thin, uniform walls to ensure consistent, precise thermal transfer for excellent results. Available with or without caps and can be ordered separately.
Brand:  BRAND™ 781280
 View more versions of this product
Product Code. 10673113
Description
Description
Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples.
  • 12-strip tube strips are available with or without caps. 12-strip caps can be ordered separately.
  • Thin, uniform walls to ensure consistent, precise thermal transfer for excellent results.
  • Autoclavable
  • RNase-, DNase-, and DNA-free.
  • Compatible with most 24-, 32-, 96-, and 384-well thermal blocks.
  • Available in convenient tube/cap sets.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Certified RNase/DNase-free
Autoclavable
Polypropylene
Yes
Strip Plate
Clear
0.2 mL
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number24151 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number781280
Lot Number23191 Certificate TypeCertificate of Analysis Date18/02/2024 Catalog Number781280
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ PCR 12-Strip Tubes > Tube strip; 12 x 0.2mL; Clear