Learn More
Cytiva PD SpinTrap™ G-25 Desalting Columns
Use for convenient and fast small-scale cleanup of proteins and other large biomolecules (>5000 Mr). Cytiva Life Sciences™ PD SpinTrap™ G-25 Desalting Columns provide the option of running multiple samples in parallel.
Brand:  Cytiva 28-9180-04
Product Code. 11783329
  • 333.00 € / Pack of 50
Edge
Explore more special offers
Description
Description
  • High recovery (70 to 90%, biomolecule dependent) for a range of applications, such as desalting, buffer exchange, and removal of low-molecular weight compounds
  • High desalting capacity (>85%); prepacked with 500μL Sephadex™ G-25 Medium
  • Reproducible, high throughput, parallel cleanup of protein samples
  • Sample preparation in the range of 100 to 180 μL without sample dilution
  • For use with standard microcentrifuge
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Range of applications such as desalting, buffer exchange, and removal of low-molecular weight compounds
≈600 μL
50 Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva PD SpinTrap™ G-25 Desalting Columns > Bed volume 500μL; 70 to 130μL sample; 50/Pk