missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ PE IBC Station with PE Pallet
The PE IBC Station is approved for the safe storage of water pollutants.
Brand: asecos™ 15480
This item is not returnable.
View return policy
Description
- Highly resistant against oil, acid, alkali, etc.
- Easy and safe in-house transport of the empty sump by forklift truck
- Ideal for storing 1,000 litre IBC or small containers
- 2,000 kg load capacity per storage area depending on the self-weight of the container
- Ideal suited for storing liquids with a maximum specific weight of up to 1,800 kg/m3
Specifications
Blue | |
1000 L | |
IBC Spill Pallet | |
2260.00 mm | |
2260.00 mm | |
745.00 mm | |
1430.00 mm | |
141 kg |
264.2 gal. | |
Polyethylene | |
88.98 in. | |
88.98 in. | |
29.33 in. | |
56.30 in. | |
310.85 lb. |