missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Plastilab™ Polypropylene Pipette Jar

Rugged jar for storing pipettes.

Brand:  Kartell™ 0021800

Product Code. 10408282

  • 170.56 € / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Rugged jar for storing pipettes.

  • Polypropylene cushioning to avoid chipped ends
  • Jar autoclavable at 120°C
  • Welded flanged base provides stability
  • Tight fitting cover for protection during transportation
Specifications

Specifications

Storing Pipette
162mm
Pipette Jar
650 mm
PP jar with PE cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kartell™ Plastilab™ Polypropylene Pipette Jar >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.