missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PE Snap Cap, Transparent, for Snap Cap Vial
Polyethylene caps are chemically inert and suitable for most chromatography applications.
Brand: Fisherbrand™ 18 08 0913
41.65 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
- PE Snap Cap
- Transparent
- Closed top
- Snap caps are easy to apply and easy to remove, eliminate the need for crimping or de-capping tools

Specifications
Transparent | |
Polyethylene | |
Snap Cap |
18 mm | |
1000 | |
ND18 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ PE Snap Cap, Transparent, for Snap Cap Vial > 18mm
Spot an opportunity for improvement?Share a Content Correction