Learn More
Lenz Laborglasinstrumente™ Pear-Shaped Flasks with Conical Ground Joint
Especially for distillation and evaporation and provides for ease of sample removal. Lenz Laborglasinstrumente™ Pear-Shaped Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 03031410
 View more versions of this product
Product Code. 10479552
  • 146.00 € / Pack of 10
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12383
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
5 mL
Borosilicate Glass, Duran Glass
14/23 NS
27 mm