Learn More
Lenz Laborglasinstrumente™ Pear-Shaped Flasks with Conical Ground Joint
Especially for distillation and evaporation and provides for ease of sample removal. Lenz Laborglasinstrumente™ Pear-Shaped Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
53.00 € - 391.00 €
Product Code | Brand | Diameter (Metric) Outer | Capacity (Metric) | Socket Joint | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Capacity (Metric) | Socket Joint | Price | Quantity & Availability | |||
10087780
|
Lenz Laborglasinstrumente™
03031437 |
64 mm | 100 mL | 14/23 NS |
103.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10508631
|
Lenz Laborglasinstrumente™
03031937 |
64 mm | 100 mL | 19/26 NS |
228.00 €
Pack of 10 |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||
10144592
|
Lenz Laborglasinstrumente™
03032937 |
64 mm | 100 mL | 29/32 NS |
391.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10000251
|
Lenz Laborglasinstrumente™
03031949 |
85 mm | 250 mL | 19/26 NS |
291.00 €
Pack of 10 |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||
10683501
|
Lenz Laborglasinstrumente™
03031422 |
42 mm | 25 mL | 14/23 NS |
65.00 €
Pack of 10 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||
10693501
|
Lenz Laborglasinstrumente™
03031428 |
51 mm | 50 mL | 14/23 NS |
84.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
11572304
|
Lenz Laborglasinstrumente™
03032928 |
51 mm | 50 mL | 29/32 NS |
358.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
10479552
|
Lenz Laborglasinstrumente™
03031410 |
27 mm | 5 mL | 14/23 NS |
146.00 €
Pack of 10 |
Estimated Shipment: 17-02-2025
Log in to see stock. |
|||
15370895
|
Lenz Laborglasinstrumente™
03031413 |
33 mm | 10 mL | 14/23 NS |
53.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||
Description
- Acc. to DIN 12383
- With inscription label
- Corrosion-free and resistant to thermal shock
- With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Borosilicate Glass, Duran Glass |