Learn More
Gilac™ Pedal Container with Lid, 50 L
Pedal Container with Lid is a compact pedal container for space saving and hands-free operation to prevent contamination.
Brand:  Gilac™ G614324
 View more versions of this product
Product Code. 17968841
  • 80.95 € / Each
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Compact pedal container for space saving.
  • Hands-free operation to prevent contamination and facilitate barrier gestures, pedal prevents contamination.
  • 2 pedal mounting positions available: automatic or manual lid closing.
  • The lid (in manual lid version) can be closed by pressing the left part of the pedal.
  • Longevity and durability.
  • Perfect stability.
  • Easy to clean.
  • Colored lids for selective waste sorting.
  • Self-assembled kit: lid, body, screws and pedal delivered in the same box.
Specifications
Specifications
50 L
Red
Polypropylene
13.2 gal.
Not Autoclavable
Pedal Container with Lid