missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Pedal Container with Lid, 50 L

Pedal Container with Lid is a compact pedal container for space saving and hands-free operation to prevent contamination.

80.95 €

Specifications

Capacity (Metric) 50 L
Capacity (English) 13.2 gal.
Autoclavable Not Autoclavable
Material Polypropylene
Product Type Pedal Container with Lid
View More Specs

Products 5
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
17988521
View Documents
Gilac™
G614321
White
80.95 €
Each
Please to purchase this item. Need a web account? Register with us today!
17908591
View Documents
Gilac™
G614322
Blue
80.95 €
Each
Please to purchase this item. Need a web account? Register with us today!
17998921
View Documents
Gilac™
G614323
Green
80.95 €
Each
Please to purchase this item. Need a web account? Register with us today!
17968841
View Documents
Gilac™
G614324
Red
80.95 €
Each
Please to purchase this item. Need a web account? Register with us today!
17948451
View Documents
Gilac™
G614325
Yellow
80.95 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Compact pedal container for space saving.
  • Hands-free operation to prevent contamination and facilitate barrier gestures, pedal prevents contamination.
  • 2 pedal mounting positions available: automatic or manual lid closing.
  • The lid (in manual lid version) can be closed by pressing the left part of the pedal.
  • Longevity and durability.
  • Perfect stability.
  • Easy to clean.
  • Colored lids for selective waste sorting.
  • Self-assembled kit: lid, body, screws and pedal delivered in the same box.
Specifications

Specifications

50 L
Not Autoclavable
Pedal Container with Lid
13.2 gal.
Polypropylene
Videos
SDS
Documents

Documents

Product Certifications