Learn More
Tocris Bioscience™ PEI STAR™ transfection reagent
Polyethylenimine (PEI) transfection reagent, chemically-defined
Brand:  Tocris Bioscience™ 7854/100
115.76 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18757876
  • 152.00 € / 100mg
Explore more special offers
Description
Description
PEI STAR™ is a chemically-defined, high-performance polyethylenimine (PEI) transfection reagent for cost-effective, affordable and scalable transient gene expression. PEI is a synthetic polymer with an exceptionally high positive charge density in pH-neutral solutions. Positively charged PEI binds strongly to negatively charged DNA and imparts a net cationic charge, allowing the DNA to enter cells. PEI is a non-viral vector commonly used to transfect HEK293 and CHO cells. Applications include production of recombinant proteins, antibodies and viruses.
PEI STAR is a trademark of Bio-Techne Corp.
Specifications
Specifications
Poly[imino(1,2-ethanediyl)] hydrochloride
49553-93-7
100 mg
Transfection Reagent