Learn More
Tocris Bioscience™ PEI STAR™ transfection reagent
Polyethylenimine (PEI) transfection reagent, chemically-defined
152.00 € - 723.00 €
Specifications
Chemical Name or Material | Poly[imino(1,2-ethanediyl)] hydrochloride |
---|---|
CAS | 49553-93-7 |
Target | Transfection Reagent |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18757876
|
Tocris Bioscience™
7854/100 |
100 mg |
152.00 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18707786
|
Tocris Bioscience™
7854/1G |
1 g |
723.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PEI STAR™ is a chemically-defined, high-performance polyethylenimine (PEI) transfection reagent for cost-effective, affordable and scalable transient gene expression. PEI is a synthetic polymer with an exceptionally high positive charge density in pH-neutral solutions. Positively charged PEI binds strongly to negatively charged DNA and imparts a net cationic charge, allowing the DNA to enter cells. PEI is a non-viral vector commonly used to transfect HEK293 and CHO cells. Applications include production of recombinant proteins, antibodies and viruses.PEI STAR is a trademark of Bio-Techne Corp.
Specifications
Poly[imino(1,2-ethanediyl)] hydrochloride | |
49553-93-7 | |
Transfection Reagent |
Safety and Handling
Recommended Storage : Store the unopened product at room temperature. Do not use past expiration date.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.