Learn More
3M™ PELTOR™ Earmuffs
PELTOR™ X2 Earmuffs are our innovative hearing protectors with a modern slim cup design.
Brand:  3M™ 7000103990
259.15 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17863276
  • 352.00 € / Pack of 10
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For high performance hearing protection against moderate noise levels, use 3M™ PELTOR™ X2 Earmuffs.
  • 3M PELTOR Earmuffs X2 are colour-coded yellow, making it simple to select the correct level of protection from the X series range.
  • The headband model features an electrically insulated, twin headband design that helps reduce heat build-up on top of the head, and the stainless steel wire, coated with plastic makes the headband the most robust 3M has to offer.
  • It is both strong and comfortable as the flat wide headband has a soft plastic coating providing stable pressure on top of the head.
  • Carefully moulded sliders ensure the earmuffs are easy to adjust when on the head.
  • Wide, soft foam ear cushions help workers stay comfortable, while the slim cups offer improved compatibility with other safety equipment.
  • Offering a product to meet each users job or preference the helmet mounted version is available as P5 with the same dielectric performance as the headband model.
  • Alternatively, if the application does not require electrical insulation the helmet mounted P3 model and neckband is available.
  • The cushions and foam inserts, available as Hygiene Kits, are easy to replace if they become worn or damaged, helping extend the life of the product.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24 dB
Oval
No
No
Hard Hat
Uncorded
Reusable
None
Hard Hat
No