missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ WS™ ProTac XPI Level Dependent Bluetooth™ Headset

3M™ PELTOR™ WS™ ProTac™ XPI is a range of level-dependent Bluetooth headsets for effective hearing protection without compromising communication.

Brand:  3M™ 7100195960

563.57 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 17838139

  • 766.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Helps provide hearing protection and wireless communication
  • Level-dependent for ambient listening
  • Bluetooth connectivity for seamless connection
  • Bluetooth MultiPoint for simultaneous connection of up to two devices
  • IP68 classified noise-cancelling boom microphone for easy inter-user communication;High visibility yellow for added safety
  • PLT (push-to-listen) feature for simple activation of level-dependent function;Available as a headband or helmet attachment
  • SNR 33 dB (SNR 31 dB with helmet attached)
  • SNR 34 dB (used in combination with HY80 gel rings, 32 dB if helmet attached
  • Uses AA batteries or rechargeable NiMH pack
  • 45 hour battery life
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Universal
Reusable
Yellow
Hardhat attached
31 to 33 dB
Yes
No
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.