Learn More
Pentafluoropropionic anhydride, 98%, Thermo Scientific Chemicals
47.10 € - 499.49 €
Chemical Identifiers
CAS | 356-42-3 |
---|---|
Molecular Formula | C6F10O3 |
Molecular Weight (g/mol) | 310.047 |
MDL Number | MFCD00000429 |
InChI Key | XETRHNFRKCNWAJ-UHFFFAOYSA-N |
Synonym | pentafluoropropionic anhydride, perfluoropropionic anhydride, pentafluoropropionic acid anhydride, pfpa, perfluoropropionic acid anhydride, propanoic acid, pentafluoro-, anhydride, unii-0lfe7u11o5, pentafluoropropanoic anhydride, decafluoropropionic anhydride, propionic acid, pentafluoro-, anhydride |
PubChem CID | 67742 |
IUPAC Name | 2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate |
SMILES | C(=O)(C(C(F)(F)F)(F)F)OC(=O)C(C(F)(F)F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11441798
|
Thermo Scientific Alfa Aesar
A14472.06 |
5 g | N/A | ||||||
11451798
|
Thermo Scientific Alfa Aesar
A14472.14 |
25 g |
177.89 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11461798
|
Thermo Scientific Alfa Aesar
A14472.22 |
100 g |
499.49 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pentafluoropropionic anhydride act as a derivatising agent of amines and alcohols. It is used in the in preparation of halogenated derivatives of phenolic acids, required for determination of some biologically important acids in cerebrospinal fluid, as derivatization reagent in detection of 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain by GC-multiple ion detection and in simultaneous determination of the major metabolites of dopamine in rat striatum by gas-liquid chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPentafluoropropionic anhydride act as a derivatising agent of amines and alcohols. It is used in the in preparation of halogenated derivatives of phenolic acids, required for determination of some biologically important acids in cerebrospinal fluid, as derivatization reagent in detection of 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain by GC-multiple ion detection and in simultaneous determination of the major metabolites of dopamine in rat striatum by gas-liquid chromatography.
Solubility
Reacts with water.
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
Chemical Identifiers
356-42-3 | |
310.047 | |
XETRHNFRKCNWAJ-UHFFFAOYSA-N | |
67742 | |
C(=O)(C(C(F)(F)F)(F)F)OC(=O)C(C(F)(F)F)(F)F |
C6F10O3 | |
MFCD00000429 | |
pentafluoropropionic anhydride, perfluoropropionic anhydride, pentafluoropropionic acid anhydride, pfpa, perfluoropropionic acid anhydride, propanoic acid, pentafluoro-, anhydride, unii-0lfe7u11o5, pentafluoropropanoic anhydride, decafluoropropionic anhydride, propionic acid, pentafluoro-, anhydride | |
2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate |
Specifications
356-42-3 | |
73°C to 74°C | |
MFCD00000429 | |
1806446 | |
pentafluoropropionic anhydride, perfluoropropionic anhydride, pentafluoropropionic acid anhydride, pfpa, perfluoropropionic acid anhydride, propanoic acid, pentafluoro-, anhydride, unii-0lfe7u11o5, pentafluoropropanoic anhydride, decafluoropropionic anhydride, propionic acid, pentafluoro-, anhydride | |
XETRHNFRKCNWAJ-UHFFFAOYSA-N | |
2,2,3,3,3-pentafluoropropanoyl 2,2,3,3,3-pentafluoropropanoate | |
67742 | |
98% |
1.589 | |
C6F10O3 | |
UN3265 | |
Moisture sensitive | |
Reacts with water. | |
C(=O)(C(C(F)(F)F)(F)F)OC(=O)C(C(F)(F)F)(F)F | |
310.047 | |
310.04 | |
Pentafluoropropionic anhydride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H332-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 206-604-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.