Learn More
Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap™ Neo Trap Cartridge to your HPLC system.
Brand:  Thermo Scientific™ 174502
 View more versions of this product
Product Code. 17437583
  • 546.11 € / Each
Estimated Shipment: 08-01-2025
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap Neo Trap Cartridge (Catalog Number 174500) to your HPLC system.
See Viper and nanoViper resources
TRUSTED_SUSTAINABILITY
Specifications
Specifications
30 μm (tubing)
Nano Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings
22,000 psi (1500 bar)
60°C
Nano Trap Cartridge Holder with PEEK Tubing and nanoViper™ Fittings
PepMap™ Neo Trap Cartridge (part number 174500)
5 mm
Each
PepMap Neo
Trap and Elute, Peptide Mapping