Learn More
Perchloric acid, 0.1N in Acetic Acid Standardized Solution, Thermo Scientific Chemicals
Description
Perchloric acid, 0.1N in acetic acid standardized solution is used in volumetric analysis. It is useful for its super acid properties and it is also used as an etchant for certain metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Appearance | Colorless liquid |
Color | Colorless |
Melting Point | 16.6°C |
Odor | Pungent |
Vapor Pressure | 16 hPa (17mm Hg) at 20°C |
Flash Point | 39°C |
Quantity | 1 L |
Physical Form | Liquid |
UN Number | 2920 |
Solubility Information | Miscible with water. |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P16L516 | Certificate of Analysis | 27/03/2025 | 035608.K7,035608.K2 |
N14L502 | Certificate of Analysis | 20/02/2025 | 035608.K7,035608.K2 |
62500064 | Certificate of Analysis | 05/02/2025 | 035608.K7,035608.K2 |
Z27K501 | Certificate of Analysis | 21/01/2025 | 035608.K7,035608.K2 |
X29K504 | Certificate of Analysis | 20/11/2024 | 035608.K7,035608.K2 |
Safety and Handling


- Perchloric acid, 0.1N in Acetic Acid Standardized Solution
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Acetic acid, Perchloric acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.