missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Percoll™ Centrifugation Media

For separation of cells, subcellular particles, and larger viruses (down to approx. 70S) under gentle conditions that preserve viability and morphological integrity. Cytiva™ Percoll™ Centrifugation Media Percoll is composed of colloidal silica coated with polyvinylpyrrolidone (PVP).

Brand:  Cytiva 17-0891-02

 View more versions of this product

Product Code. 11530734

  • 219.00 € / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Gradients can either be preformed or spontaneously generated by centrifugation at moderate speeds in an angle-head rotor
  • Low viscosity of media allows cell isolations on preformed gradients to be completed in only a few minutes using low centrifugal forces (200 to 1000xG)
  • Iso-osmotic gradients throughout cover a range of densities up to 1.3g/mL
  • Nontoxic, almost chemically inert and does not adhere to membranes
  • Adjustable to physiological ionic strength and pH; can be buffered within pH range 5.5 to 10.0 without change in properties
  • Percoll is a silica sol with nondialyzable PVP coating; 15-30nm dia. bead size

 

For in vitro research use only.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

250 mL
Cell Isolation
< 25 mOsm/kg
8.5 to 9.5
1.135 g/mL
4–30°C
15cP
Colorless to Slightly Yellow
Centrifugation Media
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Percoll™ Centrifugation Media > Percoll; PVP-coated; 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.