Learn More
Brady™ Small Adjustable Lockout Kit
The combination lockout kit gives you everything you need to lock out the most common mechanical and electrical control points.
Brand: Brady™ 806182
This item is not returnable.
View return policy
Description
- Kit for valve and electrical lockout
Specifications
Lockout/Tagout Kit | |
050979 Large Red Lockout Bag, 064057 Adjustable Case for Gate Valves, 050947 Universal Cable Lockout System + Vinyl Coated Metal Cable (2.44 m), 051339 Brady Red Safety Locks (2 x), 805840 Multi Padlock Shackles - 25 mm (2 x), 050924 Universal Lockout System - Small, 065402 Block Arm - small, 256156 Tag Do not operate 75 x 160 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Brady™ Small Adjustable Lockout Kit > Kit
Spot an opportunity for improvement?Share a Content Correction