Learn More
Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI) Acylation Reagents
Conduct acylation of hydroxyl groups and primary and secondary amines with the Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI).
722.28 €
Specifications
For Use With FID, ECD
Standard Concentration 98 to 100%
View More Specs
Products 1
Product Code Brand Standard Components Price Quantity & Availability  
Product Code Brand Standard Components Price Quantity & Availability  
11747067
View Documents
Thermo Scientific™
TS-48882
1-(Trifluoroacetyl)-1 H-imidazole This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Conduct acylation of hydroxyl groups and primary and secondary amines with the Thermo Scientific™ Perfluoroacylimidazoles (TFAI and HFBI). Compared to acid chlorides and anhydrides, the reactions of fluorinated acylimidazoles are smooth and quantitative, and produce no acid byproducts. The principal byproduct is imidazole, which is relatively inert. The volatile and stable fluoroacyl derivatives are ideal for FID and electron-capture detection (ECD) techniques.
Applications
Used in bifunctional derivatization schemes and in exchange reactions in which TMS derivatives are converted to HFB derivatives.
Hydroxyl groups of catecholamines are derivatized with TMSI, followed by conversion of the amines to acylamides with HFBI.
Tryptamine and metabolites present in spinal fluid have been analyzed by ECD using HFBI.
Specifications
Specifications
FID, ECD
98 to 100%