Learn More
Perfluorohexanes, 98+%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 355-42-0 |
---|---|
Molecular Formula | C6F14 |
Molecular Weight (g/mol) | 338.044 |
MDL Number | MFCD00000437 |
InChI Key | ZJIJAJXFLBMLCK-UHFFFAOYSA-N |
Synonym | perfluorohexane, perflexane, tetradecafluorohexane, perfluoro-n-hexane, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, fluorinert fc72, perfluorohexanes, perflexane usan:inn |
PubChem CID | 9639 |
ChEBI | CHEBI:39427 |
IUPAC Name | 1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane |
SMILES | C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11327148
|
Thermo Scientific Alfa Aesar
015812.14 |
25 g |
N/A
|
N/A | |||||
11317148
|
Thermo Scientific Alfa Aesar
015812.22 |
100 g |
N/A
|
N/A | |||||
11490480
|
Thermo Scientific Alfa Aesar
015812.36 |
500 g |
N/A
|
N/A | |||||
Description
Perfluorohexanes is used as a coolant, photosensitizer in fluorous biphasic singlet oxygenation and an inert reaction medium. It is used in the electronic cooling liquid and insulator. It plays a vital role as a reaction medium, especially for photooxidation reactions. Further, it finds application as an ultrasound contrast agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
355-42-0 | |
338.044 | |
ZJIJAJXFLBMLCK-UHFFFAOYSA-N | |
9639 | |
1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane |
C6F14 | |
MFCD00000437 | |
perfluorohexane, perflexane, tetradecafluorohexane, perfluoro-n-hexane, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, fluorinert fc72, perfluorohexanes, perflexane usan:inn | |
CHEBI:39427 | |
C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F |
Specifications
355-42-0 | |
Colorless | |
58°C to 60°C | |
C6F14 | |
1.251 | |
1802113 | |
Immiscible with water. | |
C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F | |
338.044 | |
CHEBI:39427 | |
≥98% | |
Perfluorohexanes |
-4°C | |
1.699 g/mL | |
None | |
CF3(CF2)4CF3 | |
MFCD00000437 | |
perfluorohexane, perflexane, tetradecafluorohexane, perfluoro-n-hexane, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, fluorinert fc72, perfluorohexanes, perflexane usan:inn | |
ZJIJAJXFLBMLCK-UHFFFAOYSA-N | |
1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane | |
9639 | |
338.04 | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 206-585-0
missing translation for 'rtecsNumber' : MO4310000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only