missing translation for 'onlineSavingsMsg'
Learn More
Perhydrocyclobuta[c]furan-1,3-dione, 97%, Thermo Scientific™ Product Code.: 10384412

Perhydrocyclobuta[c]furan-1,3-dione, 97%, Thermo Scientific™

Product Code. 10384412
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 10384412

Brand: Thermo Scientific Maybridge BTB13668CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4462-96-8
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
InChI Key NMNZZIMBGSGRPN-UHFFFAOYSA-N
Synonym 3-oxabicyclo 3.2.0 heptane-2,4-dione, perhydrocyclobuta c furan-1,3-dione, 1,2-cyclobutanedicarboxylic anhydride, cyclobutane-1,2-dicarboxylic anhydride, 1,2-cyclobutanedicarboxylic anhydride, cis-,, acmc-1ahmk, 3-oxabicyclo 3.2.0 heptane-2, cisShow MoreShow Less
PubChem CID 138261
IUPAC Name 3-oxabicyclo[3.2.0]heptane-2,4-dione
SMILES C1CC2C1C(=O)OC2=O

Specifications

Assay Percent Range 95%
Quantity 250 mg
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material Perhydrocyclobuta[c]furan-1, 3-dione
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Perhydrocyclobuta[c]furan-1,3-dione, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.