Learn More
Periodic acid, 99%, Thermo Scientific Chemicals
46.30 € - 528.00 €
Chemical Identifiers
CAS | 10450-60-9 |
---|---|
Molecular Formula | H5IO6 |
Molecular Weight (g/mol) | 227.94 |
MDL Number | MFCD00011344 |
InChI Key | TWLXDPFBEPBAQB-UHFFFAOYSA-N |
Synonym | orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst |
PubChem CID | 25289 |
ChEBI | CHEBI:29150 |
IUPAC Name | pentahydroxy(oxo)-$l^{7}-iodane |
SMILES | O[I](O)(O)(O)(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11444387
|
Thermo Scientific Alfa Aesar
B20433.18 |
50 g |
46.30 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454387
|
Thermo Scientific Alfa Aesar
B20433.30 |
250 g |
164.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434387
|
Thermo Scientific Alfa Aesar
B20433.0B |
1000 g |
528.00 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Periodic acid is used as a component for glycoprotein stains. It serves as a pharmaceutical and synthetic intermediate and is used in silk screen printing. It acts as an oxidizing agent especially in the cleavage of 1,2 diols to prepare dialdehydes. It has selective oxidation powers which makes it an ideal reagent for the reclamation of silk screens in the printing industry. It finds application in life sciences for the analysis of intracellular polysaccharides like glycogen, starch and cellulose. Furthermore, it is used in oxidation reactions converting sulfides into sulfones, catalyzed by chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPeriodic acid is used as a component for glycoprotein stains. It serves as a pharmaceutical and synthetic intermediate and is used in silk screen printing. It acts as an oxidizing agent especially in the cleavage of 1,2 diols to prepare dialdehydes. It has selective oxidation powers which makes it an ideal reagent for the reclamation of silk screens in the printing industry. It finds application in life sciences for the analysis of intracellular polysaccharides like glycogen, starch and cellulose. Furthermore, it is used in oxidation reactions converting sulfides into sulfones, catalyzed by chromium(VI) oxide and oxidation reactions converting alcohols to aldehydes and ketones.
Solubility
Soluble in water and alcohols.
Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong reducing agents, powdered metals, strong bases and dimethyl sulfoxide.
Chemical Identifiers
10450-60-9 | |
227.94 | |
TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
25289 | |
pentahydroxy(oxo)-$l^{7}-iodane |
H5IO6 | |
MFCD00011344 | |
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst | |
CHEBI:29150 | |
O[I](O)(O)(O)(O)=O |
Specifications
1.2 | |
126°C to 130°C | |
Hygroscopic; Light sensitive | |
MFCD00011344 | |
14,7171 | |
Soluble in water and alcohols. | |
O[I](O)(O)(O)(O)=O | |
227.94 | |
CHEBI:29150 | |
99% |
10450-60-9 | |
Odorless | |
H5IO6 | |
UN3085 | |
orthoperiodic acid, periodic acid, tetrahydroxyiodous acid, acid, periodic, paraperiodic acid, pentahydroxy-l5-iodane oxide, unii-ak1d44l87g, pentahydroxidooxidoiodine, hexaoxoiodic 5-acid, periodic acid, cryst | |
TWLXDPFBEPBAQB-UHFFFAOYSA-N | |
pentahydroxy(oxo)-$l^{7}-iodane | |
25289 | |
227.94 | |
Periodic acid |
Safety and Handling
GHS H Statement
H272-H314
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
P210-P220-P221-P260-P264b-P270-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P501c
H271-H314-H335-H372
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 233-937-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Periodic acid
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H271-May cause fire or explosion; strong oxidiser.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : periodic acid
RUO – Research Use Only