Learn More
Macherey-Nagel™ Permabond™ FFAP GC Capillary Column
Brand:  Macherey-Nagel™ 723830.25
 View more versions of this product
Product Code. 10318641
  • 646.00 € / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.5 mm
0.32 mm
G25, G35
220°C to 240°C
25 m
1 Each
GC Capillary Column
Permabond™ FFAP
Free Carboxylic Acids Analysis, Free Carboxylic Acids
0.35 μm
Permabond™ FFAP
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Permabond™ FFAP GC Capillary Column > Length: 25m; I.D.: 0.32mm; Film Thickness: 0.35um