Learn More
Macherey-Nagel™ Permabond™ FFAP GC Capillary Column
280.00 € - 1160.00 €
Specifications
Stationary Phase | Permabond™ FFAP |
---|---|
USP Type | G25, G35 |
Type | GC Capillary Column |
Temperature Range | 220°C to 240°C |
Packing Material | Permabond™ FFAP |
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Price | Quantity & Availability | ||||
10786122
|
Macherey-Nagel™
723180.10 |
10 m | 0.1 mm | 0.1 μm |
338.00 €
Each |
In Stock
Log in to see stock. |
||||
10601112
|
Macherey-Nagel™
723116.10 |
10 m | 0.25 mm | 0.25 μm |
280.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
10586501
|
Macherey-Nagel™
723181.10 |
10 m | 0.1 mm | 0.25 μm |
338.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10033011
|
Macherey-Nagel™
723830.10 |
10 m | 0.32 mm | 0.35 μm |
297.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10348641
|
Macherey-Nagel™
723344.10 |
10 m | 0.32 mm | 0.5 μm |
297.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10398301
|
Macherey-Nagel™
723555.10 |
10 m | 0.53 mm | 1 μm |
348.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10236801
|
Macherey-Nagel™
723180.20 |
20 m | 0.1 mm | 0.1 μm |
559.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10698801
|
Macherey-Nagel™
723936.25 |
25 m | 0.25 mm | 0.1 μm |
605.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10368541
|
Macherey-Nagel™
723116.25 |
25 m | 0.25 mm | 0.25 μm |
605.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10440512
|
Macherey-Nagel™
723356.25 |
25 m | 0.32 mm | 0.1 μm |
646.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10556341
|
Macherey-Nagel™
723341.25 |
25 m | 0.32 mm | 0.25 μm |
646.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10318641
|
Macherey-Nagel™
723830.25 |
25 m | 0.32 mm | 0.35 μm |
646.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10576861
|
Macherey-Nagel™
723344.25 |
25 m | 0.32 mm | 0.5 μm |
646.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10727472
|
Macherey-Nagel™
723555.25 |
25 m | 0.53 mm | 1 μm |
749.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10479931
|
Macherey-Nagel™
723116.30 |
30 m | 0.25 mm | 0.25 μm |
605.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10469741
|
Macherey-Nagel™
723341.30 |
30 m | 0.32 mm | 0.25 μm |
646.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10338021
|
Macherey-Nagel™
723344.30 |
30 m | 0.32 mm | 0.5 μm |
646.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10023011
|
Macherey-Nagel™
723116.50 |
50 m | 0.25 mm | 0.25 μm |
1005.00 €
Each |
In Stock
Log in to see stock. |
||||
10796122
|
Macherey-Nagel™
723936.50 |
50 m | 0.25 mm | 0.1 μm |
1005.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10708242
|
Macherey-Nagel™
723341.50 |
50 m | 0.32 mm | 0.25 μm |
1090.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10092851
|
Macherey-Nagel™
723830.50 |
50 m | 0.32 mm | 0.35 μm |
1090.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10348211
|
Macherey-Nagel™
723356.50 |
50 m | 0.32 mm | 0.1 μm |
1090.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10065841
|
Macherey-Nagel™
723344.50 |
50 m | 0.32 mm | 0.5 μm |
1090.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10072961
|
Macherey-Nagel™
723555.50 |
50 m | 0.53 mm | 1 μm |
1160.00 €
Each |
In Stock
Log in to see stock. |
||||
10216611
|
Macherey-Nagel™
723116.60 |
60 m | 0.25 mm | 0.25 μm |
1050.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10718242
|
Macherey-Nagel™
723341.60 |
60 m | 0.32 mm | 0.25 μm |
1155.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
10348021
|
Macherey-Nagel™
723344.60 |
60 m | 0.32 mm | 0.5 μm |
1155.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
Specifications
Permabond™ FFAP | |
GC Capillary Column | |
Permabond™ FFAP |
G25, G35 | |
220°C to 240°C | |
1 Each |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Macherey-Nagel™ Permabond™ FFAP GC Capillary Column