Learn More
Macherey-Nagel™ Permabond™ FFAP GC Capillary Column
Brand:  Macherey-Nagel™ 723181.10
 View more versions of this product
Product Code. 10586501
  • 338.00 € / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.4 mm
0.1 mm
G25, G35
220°C to 240°C
10 m
1 Each
GC Capillary Column
Permabond™ FFAP
Free Carboxylic Acids Analysis, Free Carboxylic Acids
0.25 μm
Permabond™ FFAP
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Permabond™ FFAP GC Capillary Column > Length: 10m; I.D.: 0.1mm; Film Thickness: 0.25um