Learn More
Pesticide Mix 10, SPEX CertiPrep™
Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 10 containing 15 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.
Brand:  SPEX Certiprep SPXPR-10
Product Code. 16380478
  • 231.00 € / 1mL
Explore more special offers
Description
Description
  • Pesticide Mix 9 10 containing 15 compounds in acetronitrile
  • Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial
Services
1 year
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Pesticide Mix 10, SPEX CertiPrep™ > Pesticide Mix 10 containing 15 compounds in acetronitrile