missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Streptavidin, lyophilized powder
A bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.
Brand: Molecular Probes™ 434302
410.40 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
This item is not returnable.
View return policy
Description
- A bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.
- Binding activity of streptavidin is determined to be >11 units per mg protein
- One unit of binding activity is defined as that amount of protein which binds 1 μg of biotin (Green, N.M., Biochem. J., 89:599,1963)
- Streptavidin is provided in lyophilized form in 10 mM phosphate buffered saline, pH 7.4
- Reconstitute with 1 mL of distilled or deionized water
- Store at 4°C

Specifications
Unconjugated | |
Streptavidin is provided in lyophilized form in 10 mM phosphate buffered saline, pH 7.4. Reconstitute with 1 ml of distilled or deionized water. Store at 4°C. | |
Lyophilized |
5 mg | |
Wet Ice | |
Streptavidin Conjugate (fluorescent) |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
PI434302
Version: 01/09
File size: 21 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZF389914 | Certificate of Analysis | 13/07/2024 | 434302 |
YL376578 | Certificate of Analysis | 25/01/2024 | 434302 |
YE372406 | Certificate of Analysis | 31/05/2023 | 434302 |
YA369107 | Certificate of Analysis | 27/01/2023 | 434302 |
XI364185 | Certificate of Analysis | 22/09/2022 | 434302 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Molecular Probes™ Streptavidin, lyophilized powder > 5 mg
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction