Learn More
Pesticide Mix 6, SPEX CertiPrep™
Use these Organic Certified Reference Materials for a wide range of HPLC/GC/LC applications. SPEX CertiPrep™ Pesticide Mix 6 containing 15 compounds in acetronitrile is ideal for pesticide screening, food safety and QuEChERS applications.
Brand: SPEX Certiprep SPXPR-6
Description
- Pesticide Mix 6 containing 15 compounds in acetronitrile
- Available and sold in 1mL ampules that are supplied with a pre-labeled transfer/storage vial
Services
1 year
Specifications
Fenvalerate (sanmarton) Pyridaben Tau-fluvalinate Quinoxyfen Alachlor Prowl (pendimet |
|
1 mL | |
C25H22ClNO3 C19H25ClN2OS C26H22ClF3N2O3 C15H8Cl2FNO C14H20ClNO2 C13H19N3O4 C18H19NO4 C10H12ClNO2 C17H13ClFN3O C22H23NO3 C9H7Cl3O3 C10H12N2O3S C15H22ClNO2 C6Cl5NO2 C9H8Cl3NO2S |
|
419.900 g/mol 364.933 g/mol 502.913 g/mol 308.134 g/mol 269.767 g/mol 281.308 g/mol 313.348 g/mol 213.661 g/mol 329.756 g/mol 349.4229 g/mol 269.509 g/mol 240.279 g/mol 283.794 g/mol 295.335 g/mol 300.589 g/mol |
Glass Ampule | |
99.1 99.5 95 99.7 99.2 99.4 99.5 99.5 98.4 99.4 97.5 99.5 98.9 99 99.5 |
|
100 μg/mL |
Safety and Handling
- Stock Organic Standard
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetonitrile
Your input is important to us. Please complete this form to provide feedback related to the content on this product.